Anti-S1 protein NTD HEK293 monoclonal antibody

Lieferant: ACROBIOSYSTEMS
SPD-M121-1MG SPD-M121-100UG
ACRBSPD-M121-1MGEA 2865 EUR
ACRBSPD-M121-1MG ACRBSPD-M121-100UG
Anti-S1 protein NTD HEK293 monoclonal antibody
Antikörper
Type: Primary
Antigen: S1 protein NTD
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: HEK293
Isotype: Human IgG1/kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR