Anti-Lhca3 Rabbit Polyclonal Antibody
Lieferant: Agrisera
AS01007
AGRIAS01007EA 402 EUR
AGRIAS01007
Anti-Lhca3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LHCA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, Arachis hypogaea, Colobanthus quitensis kunt bartl, Guzmania hybrid, Hordeum vulgare, Solanum lycopersicum, N. tabacum, Oryza sativa, Phaseolus vulgaris, Physcomitrella patens, Pinus strobus, Pisum sativum, Spinacia oleracea, Zea mays