Anti-Lhcb6 Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS01010
AGRIAS01010EA 329 EUR
AGRIAS01010
Anti-Lhcb6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LHCB6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chlamydomonas reinhardtii
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR