Anti-NifH Chicken Polyclonal Antibody

Lieferant: Agrisera
AS01021A
AGRIAS01021AEA 428 EUR
AGRIAS01021A
Anti-NifH Chicken Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NIFH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR