Anti-GDC-H Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS05074
AGRIAS05074EA 391 EUR
AGRIAS05074
Anti-GDC-H Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GDC-H
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR