Anti-Lhca5 Rabbit Polyclonal Antibody
Lieferant: Agrisera
AS05082
AGRIAS05082EA 402 EUR
AGRIAS05082
Anti-Lhca5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LHCA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, Arachis hypogaea, Hordeum vulgare, Oryza sativa, Phaseolus vulgaris, Pisum sativum, Spinacia oleracea, Zea mays