Anti-FtsH3/ FtsH10 Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS07204
AGRIAS07204EA 329 EUR
AGRIAS07204
Anti-FtsH3/ FtsH10 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FTSH3/10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. acetabulum, A. thaliana, Bryopsis hypnoides, Thalasiossira pseudonana, Hordeum vulgare, Synechocystis sp., Zea mays
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR