Anti-Dehydrin Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS07206
AGRIAS07206EA 299 EUR
AGRIAS07206
Anti-Dehydrin Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Dehydrin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, N. tabacum
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR