Anti-Rubisco Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS07218
AGRIAS07218EA 329 EUR
AGRIAS07218
Anti-Rubisco Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Rubisco
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Ramularia collo-cygni
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR