Anti-CCS Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS07219
AGRIAS07219EA 329 EUR
AGRIAS07219
Anti-CCS Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CCS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: full length
Host: Rabbit
Isotype:
Reactivity: Chlamydomonas reinhardtii
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR