Anti-Phytochrome A Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS07220
AGRIAS07220EA 329 EUR
AGRIAS07220
Anti-Phytochrome A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Phytochrome A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Hordeum vulgare
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR