Anti-PEPCK Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS07241
AGRIAS07241EA 329 EUR
AGRIAS07241
Anti-PEPCK Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PEPCK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. comosus, Mouse, Miscanthus giganteus, Panicum virgatum, Phaseolus vulgaris, Spartina alterniflora, Spartina patens, Zea mays
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR