Anti-H+ATPase Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS07260
AGRIAS07260EA 329 EUR
AGRIAS07260
Anti-H+ATPase Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: H+ATPase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, C. reinhardtii, Cucurbita moschata, Glycine max, H. vulgare, Kandelia obovata, Solanum lycopersicum, Medicago truncatula, N. tabacum, Noccaea caerulescens, Oryza sativa, Phalenopsis sogo Yukidian cultivar v3, Picea abies, Populus tremula, Pteris vittata, Ricinus communis, Spinacia oleracea, Zea mays
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR