Anti-PTA Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS07276
AGRIAS07276EA 449 EUR
AGRIAS07276
Anti-PTA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PTA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, Pisum sativum
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR