Anti-Lcm Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS08281
AGRIAS08281EA 329 EUR
AGRIAS08281
Anti-Lcm Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LCM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chlamydomonas reinhardtii, Spinach
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR