Anti-HSP21 Rabbit Polyclonal Antibody
Lieferant: Agrisera
AS08285
AGRIAS08285EA 329 EUR
AGRIAS08285
Anti-HSP21 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HSP21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Agave tequiliana var. Weber, A. thaliana, Cucumis sativus, Pinellia ternata, Silene vulgaris