Anti-HSP16.6 Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS08286
AGRIAS08286EA 329 EUR
AGRIAS08286
Anti-HSP16.6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HSP16.6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. tequilana, A. thaliana, Cucumis sativus l. var krak
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR