Anti-S1 Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS08309
AGRIAS08309EA 329 EUR
AGRIAS08309
Anti-S1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: S1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Ramularia collo-cygni
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR