Anti-NR Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS08310
AGRIAS08310EA 329 EUR
AGRIAS08310
Anti-NR Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Nitrate reductase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, C. reinhardtii, Diatom thalassiosira sp., H. vulgare, P. tricornutum bohlin accession pt1 8.6, Gracilaria gracilis, Red algae
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR