Anti-14-3-3C Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS08317
AGRIAS08317EA 443 EUR
AGRIAS08317
Anti-14-3-3C Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: 14-3-3 C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chlamydomonas reinhardtii, N. tabacum
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR