Anti-XTH-Xet Rabbit Polyclonal Antibody
Lieferant: Agrisera
AS08319
AGRIAS08319EA 443 EUR
AGRIAS08319
Anti-XTH-Xet Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: XTH-XET
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Hordeum vulgare, Oryza sativa