Anti-Conglutin gamma Rabbit Polyclonal Antibody
Lieferant: Agrisera
AS08335
AGRIAS08335EA 329 EUR
AGRIAS08335
Anti-Conglutin gamma Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Conglutin gamma
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human