Anti-ASY1 Mouse Monoclonal Antibody

Lieferant: Agrisera
AS08338
AGRIAS08338EA 380 EUR
AGRIAS08338
Anti-ASY1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: ASY1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: A. thaliana, Zea mays
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR