Anti-Toc75 Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS08345
AGRIAS08345EA 329 EUR
AGRIAS08345
Anti-Toc75 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TOC75
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Pisum sativum, Synechococcus and thermoSynechococcus, Synechocystis
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR