Anti-Toc75 Rabbit Polyclonal Antibody
Lieferant: Agrisera
AS08351
AGRIAS08351EA 329 EUR
AGRIAS08351
Anti-Toc75 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TOC75
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, N. tabacum, Pisum sativum, Spinacia oleracea, Synechococcus sp, ThermoSynechococcus sp., Synechocystis, Thellungiella salsuginea