Anti-Abeta Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS08357
AGRIAS08357EA 363 EUR
AGRIAS08357
Anti-Abeta Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Amyloid beta 1-42
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR