Anti-HSP70 Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS08371
AGRIAS08371EA 329 EUR
AGRIAS08371
Anti-HSP70 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HSP70
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR