Anti-N6-benzyladenosine Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS09426
AGRIAS09426EA 450 EUR
AGRIAS09426
Anti-N6-benzyladenosine Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: N6-benzyladenosine
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR