Anti-dihydrozeatin riboside Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS09436
AGRIAS09436EA 450 EUR
AGRIAS09436
Anti-dihydrozeatin riboside Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: dihydrozeatin riboside
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR