Anti-meta-topolin riboside Rabbit Polyclonal Antibody
Lieferant: Agrisera
AS09439
AGRIAS09439EA 450 EUR
AGRIAS09439
Anti-meta-topolin riboside Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: meta-topolin riboside
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen