Anti-ortho-topolin riboside Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS09447
AGRIAS09447EA 450 EUR
AGRIAS09447
Anti-ortho-topolin riboside Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ortho-topolin riboside
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR