Anti-AKINB1 Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS09460
AGRIAS09460EA 461 EUR
AGRIAS09460
Anti-AKINB1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AKINB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR