Anti-CBP80 Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS09531
AGRIAS09531EA 428 EUR
AGRIAS09531
Anti-CBP80 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CBP80
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: full length
Host: Rabbit
Isotype:
Reactivity: Human, Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR