Anti-PPI Rabbit Polyclonal Antibody (Biotin )

Lieferant: Agrisera
AS09546
AGRIAS09546EA 437 EUR
AGRIAS09546
Anti-PPI Rabbit Polyclonal Antibody (Biotin )
Antikörper
Type: Primary
Antigen: PPI
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, Zea mays
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR