Anti-Bromelain Rabbit Polyclonal Antibody (Biotin )

Lieferant: Agrisera
AS09553
AGRIAS09553EA 437 EUR
AGRIAS09553
Anti-Bromelain Rabbit Polyclonal Antibody (Biotin )
Antikörper
Type: Primary
Antigen: Bromelain
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Ananas comosus
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR