Anti-Peanut protein Chicken Polyclonal Antibody

Lieferant: Agrisera
AS09566
AGRIAS09566EA 282 EUR
AGRIAS09566
Anti-Peanut protein Chicken Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Peanut Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: A. thaliana, Oryza sativa
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR