Anti-IgG (H+L) Donkey Polyclonal Antibody (Biotin)
Lieferant: Agrisera
AS09611
AGRIAS09611EA 225 EUR
AGRIAS09611
Anti-IgG (H+L) Donkey Polyclonal Antibody (Biotin)
Antikörper
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Donkey
Isotype:
Reactivity: Rabbit