Anti-BiP Goat Polyclonal Antibody

Lieferant: Agrisera
AS09615
AGRIAS09615EA 461 EUR
AGRIAS09615
Anti-BiP Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: BIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR