Anti-IgG (H+L) Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: Agrisera
AS09629
AGRIAS09629EA 253 EUR
AGRIAS09629
Anti-IgG (H+L) Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR