Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 488)
Lieferant: Agrisera
AS09634
AGRIAS09634EA 180 EUR
AGRIAS09634
Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 488)
Antikörper
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse