Anti-Ricin Mouse Monoclonal Antibody [clone: MMH]

Lieferant: Agrisera
AS09649
AGRIAS09649EA 443 EUR
AGRIAS09649
Anti-Ricin Mouse Monoclonal Antibody [clone: MMH]
Antikörper
Type: Primary
Antigen: RICIN
Clonality: Monoclonal
Clone: MMH
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: A. thaliana
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR