Anti-IgG (H+L) Chicken Polyclonal Antibody (Biotin)

Lieferant: Agrisera
AS101026
AGRIAS101026EA 253 EUR
AGRIAS101026
Anti-IgG (H+L) Chicken Polyclonal Antibody (Biotin)
Antikörper
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR