Anti-IgM µ Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Agrisera
AS101035
AGRIAS101035EA 191 EUR
AGRIAS101035
Anti-IgM µ Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR