Anti-IgG (H+L) Chicken Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Lieferant: Agrisera
AS101063
AGRIAS101063EA 253 EUR
AGRIAS101063
Anti-IgG (H+L) Chicken Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antikörper
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope:
Host: Chicken
Isotype:
Reactivity: Goat, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR