Anti-IgG (H+L) Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: Agrisera
AS101075
AGRIAS101075EA 268 EUR
AGRIAS101075
Anti-IgG (H+L) Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype:
Reactivity: Goat, Human, Mouse, Rabbit, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR