Anti-kappa light chain Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Agrisera
AS101406
AGRIAS101406EA 253 EUR
AGRIAS101406
Anti-kappa light chain Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Secondary
Antigen: kappa light chain
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR