Anti-kappa light chain Goat Polyclonal Antibody (Biotin)

Lieferant: Agrisera
AS101411
AGRIAS101411EA 298 EUR
AGRIAS101411
Anti-kappa light chain Goat Polyclonal Antibody (Biotin)
Antikörper
Type: Secondary
Antigen: kappa light chain
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR