Anti-Biotin Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: Agrisera
AS101452
AGRIAS101452EA 240 EUR
AGRIAS101452
Anti-Biotin Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Type: Secondary
Antigen: Biotin
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR