Anti-IgG (H+L) Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS101492
AGRIAS101492EA 195 EUR
AGRIAS101492
Anti-IgG (H+L) Rabbit Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR