Anti-IgM µ Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS101506
AGRIAS101506EA 240 EUR
AGRIAS101506
Anti-IgM µ Rabbit Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR